1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane, 97%, Thermo Scientific Chemicals
1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number B21405.04
also known as B21405-04
Price (USD)/ Each
54.80
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
54.80
Add to cart
1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane, 97%, Thermo Scientific Chemicals
Catalog numberB21405.04
Price (USD)/ Each
54.80
-
Add to cart
Chemical Identifiers
CAS371-83-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3985-1.4035 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only