Thermo Scientific Chemicals

Tetracycline hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: B21408.22
100 g, Each
Thermo Scientific Chemicals

Tetracycline hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: B21408.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B21408.22
also known as B21408-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64-75-5
IUPAC Name
hydrogen (2Z,4S,4aS,5aS,6S,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-6,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Molecular Formula
C22H25ClN2O8
InChI Key
DYQBMRZOOXYHIM-NCKOGSTASA-N
SMILES
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)C4=C(O)C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥95.0% (non-U.S. sourced material)
Assay from Supplier's CofA
≥900 µg/mg (U.S. sourced material)
Comment
Material Sourced in the U.S. and in other countries

Description

Tetracycline hydrochloride is used to induce apoptosis in osteoclasts. It is used to treat acne and other skin infections, respiratory tract infections like pneumonia, genital, urinary infections, leptospirosis, helicobacter pylori, taxoplasmosis, mycoplasma, psittacosis for dog and cats. It also works more effectively in animals that have tick-borne infections. It is also useful in cell culture applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Tetracycline hydrochloride is the hydrochloride salt of
the antibiotic tetracycline
• Tetracycline blocks amino acyl-tRNA binding during
bacterial protein synthesis
• Tetracycline is a broad-spectrum naphthacene antibiotic that
inhibits growth of both gram positive and gram negative bacteria
Applications
• Tetracycline may be used as a selection agent in cell
culture
• It may also be used to activate inducible gene
expression in engineered cells
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text