Thermo Scientific Chemicals

Di-n-propyl ether, 99%, Thermo Scientific Chemicals

Catalog number: B21423.22
100 g, Each
Thermo Scientific Chemicals

Di-n-propyl ether, 99%, Thermo Scientific Chemicals

Catalog number: B21423.22
100 g, Each
Quantity
Catalog number: B21423.22
also known as B21423-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-43-3
IUPAC Name
1-propoxypropane
Molecular Formula
C6H14O
InChI Key
POLCUAVZOMRGSN-UHFFFAOYSA-N
SMILES
CCCOCCC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.3790-1.3820 @ 20?C

Description

Di-n-propyl ether, is used as a solvent and intermediate for organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-propyl ether, is used as a solvent and intermediate for organic chemical synthesis.

Solubility
It is slightly soluble in water 3 g/L (20°C), soluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids, halogens.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text