Diethyl 1-tetradecylphosphonate, 97+%, Thermo Scientific Chemicals
Diethyl 1-tetradecylphosphonate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl 1-tetradecylphosphonate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21426.06
also known as B21426-06
Price (USD)/ Each
84.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
84.30
Add to cart
Diethyl 1-tetradecylphosphonate, 97+%, Thermo Scientific Chemicals
Catalog numberB21426.06
Price (USD)/ Each
84.30
-
Add to cart
Chemical Identifiers
CAS5191-09-3
IUPAC Namediethyl tetradecylphosphonate
Molecular FormulaC18H39O3P
InChI KeyCPWIYYOBPHXJIA-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCP(=O)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4425-1.4465 @ 20°C
FormLiquid
Assay (GC)≥97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only