1,1-Diphenyl-2-propyn-1-ol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1-Diphenyl-2-propyn-1-ol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in medicine.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
- P Crochet.; MA Esteruelas.; E Gutiérrez-Puebla. Unusual Activation of 1, 1-Diphenyl-2-propyn-1-ol Mediated by the Os (η5-C5H5) Unit. Organometallics. 199817 (15), 3141-3142.
- E Negishi.; M Hata.; C Xu. A Strictly Pair-Selective Synthesis of Conjugated Diynes via Pd-Catalyzed Cross Coupling of 1,3-Diynylzincs: A Superior Alternative to the Cadiot-Chodkiewicz Reaction. Organic letters. 20002 (23), 3687-3689.