Thermo Scientific Chemicals

Indane, 95%, Thermo Scientific Chemicals

Catalog number: B21442.AE
100 mL, Each
Thermo Scientific Chemicals

Indane, 95%, Thermo Scientific Chemicals

Catalog number: B21442.AE
100 mL, Each
Quantity
Catalog number: B21442.AE
also known as B21442-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
496-11-7
IUPAC Name
2,3-dihydro-1H-indene
Molecular Formula
C9H10
InChI Key
PQNFLJBBNBOBRQ-UHFFFAOYSA-N
SMILES
C1CC2=CC=CC=C2C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (unspecified)
>94.0%
Refractive Index
1.5340-1.5400 @ 20?C

Description

Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.

Solubility
Soluble in ethanol, diethyl ether, chloroform (slightly), and ether. Insoluble in water.

Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text