Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.
Solubility
Soluble in ethanol, diethyl ether, chloroform (slightly), and ether. Insoluble in water.
Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from strong oxidizing agents.
RUO – Research Use Only
A Ramazani.; A Bodaghi. One-pot, four-component synthesis of dialkyl [indane-1, 3-dione-2-ylidene] alkoxysuccinates. Tetrahedron Letters. 200041 (4), 567-568.
MP Cava.; RL Litle.; DR Napier. Condensed cyclobutane aromatic systems. V. The synthesis of some α-diazoindanones: ring contraction in the indane series. J. Am. Chem. Soc. 195880 (9), 2257-2263.