Chemical Identifiers
CAS68971-82-4
IUPAC Name5,11,17,23,29,35,41,47-octa-tert-butylnonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
Molecular FormulaC88H112O8
InChI KeyOLZFZIXORGGLLS-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC8=C(O)C(CC9=C(O)C(C2)=CC(=C9)C(C)(C)C)=CC(=C8)C(C)(C)C)=CC(=C7)C(C)(C)C)=CC(=C6)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
View more