L-(-)-Galactose, 98%
L-(-)-Galactose, 98%
Thermo Scientific Chemicals

L-(-)-Galactose, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number B21448.MC
also known as B21448-MC
Price (USD)/ Each
109.00
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)/ Each
109.00
Add to cart
L-(-)-Galactose, 98%
Catalog numberB21448.MC
Price (USD)/ Each
109.00
-
Add to cart
Chemical Identifiers
CAS15572-79-9
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-78.0 ? 2.0? (c=5 in water)
Assay (unspecified)>97.5%
Appearance (Color)White
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only