Thermo Scientific Chemicals

1-Methylpyrrole, 99%, Thermo Scientific Chemicals

Catalog number: B21451.AP
500 mL, Each
Thermo Scientific Chemicals

1-Methylpyrrole, 99%, Thermo Scientific Chemicals

Catalog number: B21451.AP
500 mL, Each
Quantity
Catalog number: B21451.AP
also known as B21451-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-54-8
IUPAC Name
1-methyl-1H-pyrrole
Molecular Formula
C5H7N
InChI Key
OXHNLMTVIGZXSG-UHFFFAOYSA-N
SMILES
CN1C=CC=C1
Form
Liquid
Appearance (Color)
Clear colorless to yellow to brown
Refractive Index
1.4855-1.4885 @ 20?C
Assay (GC)
≥98.5%

Description

1-Methylpyrrole, can be used as organic synthetic materials, widely used in medicine. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpyrrole, can be used as organic synthetic materials, widely used in medicine. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in organic solvents such as alcohol and so on, but insoluble in water.

Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Stable under normal temperatures and pressures.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text