Thermo Scientific Chemicals

3-Methylanisole, 99%, Thermo Scientific Chemicals

Catalog number: B21455.30
250 g, Each
Thermo Scientific Chemicals

3-Methylanisole, 99%, Thermo Scientific Chemicals

Catalog number: B21455.30
250 g, Each
Quantity
Catalog number: B21455.30
also known as B21455-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-84-5
IUPAC Name
1-methoxy-3-methylbenzene
Molecular Formula
C8H10O
InChI Key
OSIGJGFTADMDOB-UHFFFAOYSA-N
SMILES
COC1=CC=CC(C)=C1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5120-1.5150 @ 20?C
Form
Liquid

Description

3-Methylanisole, is widely used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photo initiators and agrochemicals. It is also used as a solvent and in perfumery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylanisole, is widely used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photo initiators and agrochemicals. It is also used as a solvent and in perfumery.

Solubility
Soluble in alcohol and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text