Thermo Scientific Chemicals

Perfluoro(allylbenzene), 97%, Thermo Scientific Chemicals

Catalog number: B21469.06
5 g, Each
Thermo Scientific Chemicals

Perfluoro(allylbenzene), 97%, Thermo Scientific Chemicals

Catalog number: B21469.06
5 g, Each
Quantity
Catalog number: B21469.06
also known as B21469-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67899-41-6
IUPAC Name
1,2,3,4,5-pentafluoro-6-(1,1,2,3,3-pentafluoroprop-2-en-1-yl)benzene
Molecular Formula
C9F10
InChI Key
WRHBYJDZKRNITP-UHFFFAOYSA-N
SMILES
FC(F)=C(F)C(F)(F)C1=C(F)C(F)=C(F)C(F)=C1F
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.3820-1.3870 @ 20?C

Description

Perfluoro(allylbenzene) is used as a pharmaceutical intermediate and as a chemical in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(allylbenzene) is used as a pharmaceutical intermediate and as a chemical in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, ammonia.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text