Thermo Scientific Chemicals

1,4-Bis(2-hydroxyisopropyl)benzene, 97%, Thermo Scientific Chemicals

Catalog number: B21472.03
1 g, Each
Thermo Scientific Chemicals

1,4-Bis(2-hydroxyisopropyl)benzene, 97%, Thermo Scientific Chemicals

Catalog number: B21472.03
1 g, Each
Quantity
Catalog number: B21472.03
also known as B21472-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2948-46-1
IUPAC Name
2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol
Molecular Formula
C12H18O2
InChI Key
LEARFTRDZQQTDN-UHFFFAOYSA-N
SMILES
CC(C)(O)C1=CC=C(C=C1)C(C)(C)O
Assay (GC)
≥96.0%
Melting Point (clear melt)
137.0-146.0?C
Appearance (Color)
White to pale cream to cream to yellow to brown
Form
Powder

Description

1,4-Bis(2-hydroxyisopropyl)benzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(2-hydroxyisopropyl)benzene is used as a pharmaceutical intermediate.

Solubility
Slightly Soluble in water (2.1 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text