1,4-Bis(2-hydroxyisopropyl)benzene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Bis(2-hydroxyisopropyl)benzene is used as a pharmaceutical intermediate.
Solubility
Slightly Soluble in water (2.1 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
RUO – Research Use Only
WE Bissinger.; RH Fredenburg. Some Reactions of Butadiene Monochlorohydrin, 1-Chloro-3-buten-2-ol. J. Am. Chem. Soc. 194769 (12), 2955-2961.
C Paulo.; JE Puskas. Synthesis of hyperbranched polyisobutylenes by inimer-type living polymerization. 1. Investigation of the effect of reaction conditions. Macromolecules. 200134 (4), 734-739.