10-Bromo-1-decanol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
10-Bromo-1-decanol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
It is miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only
X Yu.; X Tang.; C Pan. Synthesis, characterization and self-assembly behavior of six-armed star block copolymers with triphenylene core. Polymer. 200546 (24), 11149-11156.
R Lu.; Y Kamiya.; T Miyakoshi. Preparation and characterization of Melanorrhoea usitata lacquer film based on pyrolysis-gas chromatography/mass spectrometry. Journal of analytical and applied pyrolysis. 200778 (1), 172-179.