2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.
Solubility
Soluble in toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light sensitive.
RUO – Research Use Only
General References:
- A Iuliano.; P Piccioli.; D Fabbri. Ring-Closing Olefin Metathesis of 2, 2'-Divinylbiphenyls: A Novel and General Approach to Phenanthrenes. Organic letters. 20046 (21), 3711-3714.
- FM Beringer.; SJ Huang . Effects of Substituents on the Rearrangement of 2-Aryl Iodoniobenzoates and Their Cleavage to Benzyne1, 2. The Journal of Organic Chemistry. 19646 (21), 3711-3714.