Diethyl phenyl orthoformate, 97%, Thermo Scientific Chemicals
Diethyl phenyl orthoformate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl phenyl orthoformate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B21498.09
also known as B21498-09
Price (USD)/ Each
52.80
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
52.80
Add to cart
Diethyl phenyl orthoformate, 97%, Thermo Scientific Chemicals
Catalog numberB21498.09
Price (USD)/ Each
52.80
-
Add to cart
Chemical Identifiers
CAS14444-77-0
IUPAC Name(diethoxymethoxy)benzene
Molecular FormulaC11H16O3
InChI KeyQTURWMMVIIBRRP-UHFFFAOYSA-N
SMILESCCOC(OCC)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4770-1.4820 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only