3-Hydroxy-4-methylbenzoic acid, 98%, Thermo Scientific Chemicals
3-Hydroxy-4-methylbenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxy-4-methylbenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number B21501.14
also known as B21501-14
Price (USD)/ Each
466.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
466.00
Add to cart
3-Hydroxy-4-methylbenzoic acid, 98%, Thermo Scientific Chemicals
Catalog numberB21501.14
Price (USD)/ Each
466.00
-
Add to cart
Chemical Identifiers
CAS586-30-1
IUPAC Name3-hydroxy-4-methylbenzoic acid
Molecular FormulaC8H8O3
InChI KeyZQLCWPXBHUALQC-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)208.0-215.0?C
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only