Thermo Scientific Chemicals

4,4,4-Trifluoro-2-butanone, 97%, Thermo Scientific Chemicals

Catalog number: B21514.06
5 g, Each
Thermo Scientific Chemicals

4,4,4-Trifluoro-2-butanone, 97%, Thermo Scientific Chemicals

Catalog number: B21514.06
5 g, Each
Quantity
Catalog number: B21514.06
also known as B21514-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2366-70-3
IUPAC Name
4,4,4-trifluorobutan-2-one
Molecular Formula
C4H5F3O
InChI Key
BTXXTMOWISPQSJ-UHFFFAOYSA-N
SMILES
CC(=O)CC(F)(F)F
Refractive Index
1.3280-1.3330 @ 20°C
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%

Description

It is used as a pharmaceutical intermediate. It is used for the synthesis of racemic 2-amino-5,5,5-trifluoro-3-methylpentanoic acid as fluorinated synthon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. It is used for the synthesis of racemic 2-amino-5,5,5-trifluoro-3-methylpentanoic acid as fluorinated synthon.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges. Keep ignition sources away. Prevent formation of aerosols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text