Thermo Scientific Chemicals

4-tert-Butylcalix[4]arene, 99%, Thermo Scientific Chemicals

Catalog number: B21515.14
25 g, Each
Thermo Scientific Chemicals

4-tert-Butylcalix[4]arene, 99%, Thermo Scientific Chemicals

Catalog number: B21515.14
25 g, Each
Quantity
Catalog number: B21515.14
also known as B21515-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60705-62-6
IUPAC Name
5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
Molecular Formula
C44H56O4
InChI Key
NVKLTRSBZLYZHK-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or flakes
Purity (DSC)
≥98.5%
Melting Point (DSC)
338-344?C
Identification (FTIR)
Conforms

Description

It is used as a starting material for further derivatization, including halogenation, acylation, and diazo coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a starting material for further derivatization, including halogenation, acylation, and diazo coupling.

Solubility
Soluble in chloroform, benzene, toluene. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text