3,3-Dimethyloctane, 98%, Thermo Scientific Chemicals
3,3-Dimethyloctane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Dimethyloctane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B21520.06
also known as B21520-06
Price (USD)/ Each
138.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
138.00
Add to cart
3,3-Dimethyloctane, 98%, Thermo Scientific Chemicals
Catalog numberB21520.06
Price (USD)/ Each
138.00
-
Add to cart
Chemical Identifiers
CAS4110-44-5
IUPAC Name3,3-dimethyloctane
Molecular FormulaC10H22
InChI KeyDBULLUBYDONGLT-UHFFFAOYSA-N
SMILESCCCCCC(C)(C)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4125-1.4165 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only