Cyclohexyl hexanoate, 99%, Thermo Scientific Chemicals
Cyclohexyl hexanoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexyl hexanoate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number B21547.22
also known as B21547-22
Price (USD)/ Each
145.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
145.00
Add to cart
Cyclohexyl hexanoate, 99%, Thermo Scientific Chemicals
Catalog numberB21547.22
Price (USD)/ Each
145.00
-
Add to cart
Chemical Identifiers
CAS6243-10-3
IUPAC Namecyclohexyl hexanoate
Molecular FormulaC12H22O2
InChI KeyDTNOERNOMHQUCN-UHFFFAOYSA-N
SMILESCCCCCC(=O)OC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.4445-1.4475 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only