4-(Chloroacetyl)morpholine, 97+%, Thermo Scientific Chemicals
4-(Chloroacetyl)morpholine, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Chloroacetyl)morpholine, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21548.03
also known as B21548-03
Price (USD)/ Each
34.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
34.50
Add to cart
4-(Chloroacetyl)morpholine, 97+%, Thermo Scientific Chemicals
Catalog numberB21548.03
Price (USD)/ Each
34.50
-
Add to cart
Chemical Identifiers
IUPAC Name2-chloro-1-(morpholin-4-yl)ethan-1-one
Molecular FormulaC6H10ClNO2
InChI KeyYMQRPXBBBOXHNZ-UHFFFAOYSA-N
SMILESClCC(=O)N1CCOCC1
Molecular Weight (g/mol)163.60
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)27.5-33.5?C
Appearance (Color)White to pale yellow or pale cream
Assay (GC)≥97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only