Thermo Scientific Chemicals

(1R)-(-)-Camphor-10-sulfonic acid, 98%, Thermo Scientific Chemicals

Catalog number: B21553.22
100 g, Each
Thermo Scientific Chemicals

(1R)-(-)-Camphor-10-sulfonic acid, 98%, Thermo Scientific Chemicals

Catalog number: B21553.22
100 g, Each
Quantity
Catalog number: B21553.22
also known as B21553-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35963-20-3
IUPAC Name
[(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid
Molecular Formula
C10H16O4S
InChI Key
MIOPJNTWMNEORI-MHPPCMCBSA-N
SMILES
CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Optical Rotation
-21.0? ? 2.5? (c=20 in water)
Appearance (Color)
White to cream

Description

(1R)-(-)-Camphor-10-sulfonic acid, is used as a pharamaceutical intermediate and also used as a chiral derivative of Camp. It can also be used as resolving agents for chiral amines and other cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R)-(-)-Camphor-10-sulfonic acid, is used as a pharamaceutical intermediate and also used as a chiral derivative of Camp. It can also be used as resolving agents for chiral amines and other cations.

Solubility
Soluble in water, and a wide variety of organic substances.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text