Anisoin, 97%, Thermo Scientific Chemicals
Anisoin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Anisoin, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Catalog number B21559.36
also known as B21559-36
Price (USD)/ Each
481.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
481.00
Add to cart
Anisoin, 97%, Thermo Scientific Chemicals
Catalog numberB21559.36
Price (USD)/ Each
481.00
-
Add to cart
Chemical Identifiers
CAS119-52-8
IUPAC Name2-hydroxy-1,2-bis(4-methoxyphenyl)ethan-1-one
Molecular FormulaC16H16O4
InChI KeyLRRQSCPPOIUNGX-UHFFFAOYNA-N
SMILESCOC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)108-115?C
Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.

Solubility
Soluble in water, ethanol and acetone.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
RUO – Research Use Only

General References:

  1. Chen, M.; Li, L.; Nie, H.; Tong, J.; Yan, L.; Xu, B.; Sun, J. S.; Tian, W.; Zhao, Z.; Qin, A.; Tang, B. Z. Tetraphenylpyrazine-based AIEgens: facile preparation and tunable light emission. Chem. Sci. 2015, 6, 1932-1937.
  2. Kamal, A.; Babu, K. S.; Hussaini, S. M. A.; Mahesh, R.; Alarifi, A. Amberlite IR-120H, an efficient and recyclable solid phase catalyst for the synthesis of quinoxalines: a greener approach. Tetrahedron Lett. 2015, 56 (21), 2803-2808.