Thermo Scientific Chemicals

2-Chloro-4-nitrophenol, 97%, Thermo Scientific Chemicals

Catalog number: B21561.30
250 g, Each
Thermo Scientific Chemicals

2-Chloro-4-nitrophenol, 97%, Thermo Scientific Chemicals

Catalog number: B21561.30
250 g, Each
Quantity
Catalog number: B21561.30
also known as B21561-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
619-08-9
IUPAC Name
2-chloro-4-nitrophenol
Molecular Formula
C6H4ClNO3
InChI Key
BOFRXDMCQRTGII-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1Cl)[N+]([O-])=O
Appearance (Color)
White to pale cream to pale yellow to pale brown to gray
Assay (GC)
≥96.0%
Melting Point (clear melt)
107.0-116.0?C
Form
Powder

Description

2-Chloro-4-nitrophenol, is frequently used as building blocks for dyes, plastics, explosives. It acts as a catalytic agent, petrochemical additive and used in organic synthesis. It can react with sulfuric acid dimethyl ester to produce 2-chloro-4-nitro-anisole. This reaction will need reagents K2CO3 and xylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-nitrophenol, is frequently used as building blocks for dyes, plastics, explosives. It acts as a catalytic agent, petrochemical additive and used in organic synthesis. It can react with sulfuric acid dimethyl ester to produce 2-chloro-4-nitro-anisole. This reaction will need reagents K2CO3 and xylene.

Solubility
Slightly Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text