It is used in the analysis of perfluorooctanesulfonate and related fluorochemicals in water and biological tissue samples by liquid chromatography-ion trap mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the analysis of perfluorooctanesulfonate and related fluorochemicals in water and biological tissue samples by liquid chromatography-ion trap mass spectrometry.
Solubility
Partly soluble in water.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Anthony A. Ribeiro and Keiko Umayahara. 19F, 13C NMR analysis of an oxygen carrier, perfluorotributylamine, and perfluoropentanoic acid.Magn. Reson. Chem.,200341(2), 107-114.
James D. Pearson, et al. Perfluorinated acid alternatives to trifluoroacetic acid for reversed-phase high-performance liquid chromatography.J. Chromatogr.. A.,1996746(2), 277-281.