Thermo Scientific Chemicals

4-Ethylresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: B21575.22
100 g, Each
Thermo Scientific Chemicals

4-Ethylresorcinol, 98%, Thermo Scientific Chemicals

Catalog number: B21575.22
100 g, Each
Quantity
Catalog number: B21575.22
also known as B21575-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2896-60-8
IUPAC Name
4-ethylbenzene-1,3-diol
Molecular Formula
C8H10O2
InChI Key
VGMJYYDKPUPTID-UHFFFAOYSA-N
SMILES
CCC1=CC=C(O)C=C1O
Assay (GC)
≥97.5%
Appearance (Color)
White to pale brown
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
94.0-100.0?C

Description

It is employed as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Solubility in methanol; almost transparency. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text