Thermo Scientific Chemicals

2,2,3,3-Tetrafluoro-1-propanol, 97%, Thermo Scientific Chemicals

Catalog number: B21594.18
50 g, Each
Thermo Scientific Chemicals

2,2,3,3-Tetrafluoro-1-propanol, 97%, Thermo Scientific Chemicals

Catalog number: B21594.18
50 g, Each
Quantity
Catalog number: B21594.18
also known as B21594-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-37-9
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.3190-1.3240 @ 20?C

Description

2,2,3,3-Tetrafluoro-1-propanol, is used as an essential fluorinated fine chemical, the main application of TFP is as a dye solvent in CD-R(W) and DVD-R(W) industry. It is also used in making textile treatment agent and pharmaceuticals. This chemical is an ideal replacement for Freon 22 detergent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluoro-1-propanol, is used as an essential fluorinated fine chemical, the main application of TFP is as a dye solvent in CD-R(W) and DVD-R(W) industry. It is also used in making textile treatment agent and pharmaceuticals. This chemical is an ideal replacement for Freon 22 detergent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text