2,2,3,3-Tetrafluoro-1-propanol, 97%, Thermo Scientific Chemicals
2,2,3,3-Tetrafluoro-1-propanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,3,3-Tetrafluoro-1-propanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number B21594.30
also known as B21594-30
Price (USD)/ Each
394.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
394.00
Add to cart
2,2,3,3-Tetrafluoro-1-propanol, 97%, Thermo Scientific Chemicals
Catalog numberB21594.30
Price (USD)/ Each
394.00
-
Add to cart
Chemical Identifiers
CAS76-37-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3190-1.3240 @ 20?C
2,2,3,3-Tetrafluoro-1-propanol, is used as an essential fluorinated fine chemical, the main application of TFP is as a dye solvent in CD-R(W) and DVD-R(W) industry. It is also used in making textile treatment agent and pharmaceuticals. This chemical is an ideal replacement for Freon 22 detergent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluoro-1-propanol, is used as an essential fluorinated fine chemical, the main application of TFP is as a dye solvent in CD-R(W) and DVD-R(W) industry. It is also used in making textile treatment agent and pharmaceuticals. This chemical is an ideal replacement for Freon 22 detergent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

General References:

  1. YJ Shih.; WN Putra.; YH Huang.; JC Tsai. Mineralization and deflourization of 2, 2, 3, 3-tetrafluoro-1-propanol (TFP) by UV/persulfate oxidation and sequential adsorption. Chemosphere. 201289 (10), 1262-1266.
  2. CS Tan.; PL Lee. Supercritical CO2 desorption of activated carbon loaded with 2, 2, 3, 3-tetrafluoro-1-propanolin a rotating packed bed. Environmental science & technology. 200842 (6), 2150-2154.