Thermo Scientific Chemicals

1-Iodo-3-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: B21605.22
100 g, Each
Thermo Scientific Chemicals

1-Iodo-3-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: B21605.22
100 g, Each
Quantity
Catalog number: B21605.22
also known as B21605-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
645-00-1
IUPAC Name
1-iodo-3-nitrobenzene
Molecular Formula
C6H4INO2
InChI Key
CBYAZOKPJYBCHE-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC(I)=C1
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥98.5%
Appearance (Color)
White to pale cream to cream to brown
Melting Point (clear melt)
32.0-39.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text