Thermo Scientific Chemicals

2-Thioxanthine, 98+%, Thermo Scientific Chemicals

Catalog number: B21606.03
1 g, Each
Thermo Scientific Chemicals

2-Thioxanthine, 98+%, Thermo Scientific Chemicals

Catalog number: B21606.03
1 g, Each
Quantity
Catalog number: B21606.03
also known as B21606-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2487-40-3
IUPAC Name
2-sulfanylidene-2,3,6,7-tetrahydro-1H-purin-6-one
Molecular Formula
C5H4N4OS
InChI Key
XNHFAGRBSMMFKL-UHFFFAOYSA-N
SMILES
O=C1NC(=S)NC2=C1NC=N2
Appearance (Color)
Yellow
Form
Powder
Assay (HPLC)
≥98.0%

Description

2-Thioxanthine used to inhibit MPO (Myeloperoxidase).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thioxanthine used to inhibit MPO (Myeloperoxidase).

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text