Thermo Scientific Chemicals

5-Bromo-m-xylene, 98%, Thermo Scientific Chemicals

Catalog number: B21610.14
25 g, Each
Thermo Scientific Chemicals

5-Bromo-m-xylene, 98%, Thermo Scientific Chemicals

Catalog number: B21610.14
25 g, Each
Quantity
Catalog number: B21610.14
also known as B21610-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
556-96-7
IUPAC Name
1-bromo-3,5-dimethylbenzene
Molecular Formula
C8H9Br
InChI Key
LMFRTSBQRLSJHC-UHFFFAOYSA-N
SMILES
CC1=CC(Br)=CC(C)=C1
Refractive Index
1.5470-1.5510 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

5-Bromo-m-xylene is used in the preparation of substituted 2,2'-bis(diphenylphosphanylmethyl)-1,1'-binaphthyl derivatives. It is also used to prepare o-tolyl-3,5-xylyl ether by reacting with o-cresol. Further, it is an organic building block used in the synthesis of various chemicals as well as in the preparation of tri-o-tolylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-m-xylene is used in the preparation of substituted 2,2′-bis(diphenylphosphanylmethyl)-1,1′-binaphthyl derivatives. It is also used to prepare o-tolyl-3,5-xylyl ether by reacting with o-cresol. Further, it is an organic building block used in the synthesis of various chemicals as well as in the preparation of tri-o-tolylphosphine.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text