L-(-)-Xylose, 99%
Thermo Scientific Chemicals

L-(-)-Xylose, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B21622.14
also known as B21622-14
Price (USD)/ Each
165.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
165.00
Add to cart
L-(-)-Xylose, 99%
Catalog numberB21622.14
Price (USD)/ Each
165.00
-
Add to cart
Chemical Identifiers
CAS609-06-3
IUPAC Name2,3,4,5-tetrahydroxypentanal
Molecular FormulaC5H10O5
InChI KeyPYMYPHUHKUWMLA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-18° to -20° (c=10 in water)
Assay from Suppliers CofA≥98.5% (Unspecified)
Water Content (Karl Fischer Titration)≤0.5%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only