Thermo Scientific Chemicals

Ethylhydrocupreine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: B21627.MC
100 mg, Each
Thermo Scientific Chemicals

Ethylhydrocupreine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: B21627.MC
100 mg, Each
Quantity
Catalog number: B21627.MC
also known as B21627-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3413-58-9
IUPAC Name
hydrogen (R)-(6-ethoxyquinolin-4-yl)[(2S,5R)-5-ethyl-2,5-dimethyl-1-azabicyclo[2.2.2]octan-2-yl]methanol chloride
Molecular Formula
C23H33ClN2O2
InChI Key
UBGKQKTVBOWPKR-BVMFEKKJSA-N
SMILES
[H+].[Cl-].CCOC1=CC=C2N=CC=C([C@@H](O)[C@]3(C)CC4CCN3C[C@]4(C)CC)C2=C1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms
Optical Rotation
-120° to -130° (c=1 in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text