Thermo Scientific Chemicals

Tartronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21640.77
0.1 g, Each
Thermo Scientific Chemicals

Tartronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21640.77
0.1 g, Each
Quantity
Catalog number: B21640.77
also known as B21640-77
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80-69-3
IUPAC Name
2-hydroxypropanedioic acid
Molecular Formula
C3H4O5
InChI Key
ROBFUDYVXSDBQM-UHFFFAOYSA-N
SMILES
OC(C(O)=O)C(O)=O
Appearance (Color)
White
Assay from Suppliers CofA
≥96.0% (Proton NMR)
Proton NMR
Conforms to structure
Form
Crystals or powder or crystalline powder

Description

Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.

Solubility
Soluble in water and alcohol.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text