Thermo Scientific Chemicals

Isobutyramide, 99%, Thermo Scientific Chemicals

Catalog number: B21641.36
500 g, Each
Thermo Scientific Chemicals

Isobutyramide, 99%, Thermo Scientific Chemicals

Catalog number: B21641.36
500 g, Each
Quantity
Catalog number: B21641.36
also known as B21641-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
563-83-7
IUPAC Name
2-methylpropanamide
Molecular Formula
C4H9NO
InChI Key
WFKAJVHLWXSISD-UHFFFAOYSA-N
SMILES
CC(C)C(N)=O
Assay (GC)
≥98.5%
Appearance (Color)
White
Melting Point (clear melt)
126-130?C
Form
Crystals or powder or crystalline powder or flakes
Identification (FTIR)
Conforms

Description

Isobutyramide plays an important role in chemical grafting of human serum albumin during the synthesis of sequentially assembled protein capsules. It is used in pharmaceutical manufacturing and in cellular differentiation. It finds application in the transcription of human gamma-globin gene and murine embryonic epsilon(y)-globin gene. Further, it is in the treatment of beta-thalassemia and sickle cell disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with acids, bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text