Thermo Scientific Chemicals

3-(2-Furyl)acrylic acid, 99%, Thermo Scientific Chemicals

Catalog number: B21653.30
250 g, Each
Thermo Scientific Chemicals

3-(2-Furyl)acrylic acid, 99%, Thermo Scientific Chemicals

Catalog number: B21653.30
250 g, Each
Quantity
Catalog number: B21653.30
also known as B21653-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
539-47-9
IUPAC Name
(2E)-3-(furan-2-yl)prop-2-enoic acid
Molecular Formula
C7H6O3
InChI Key
ZCJLOOJRNPHKAV-ONEGZZNKSA-N
SMILES
OC(=O)C=CC1=CC=CO1
Appearance (Color)
Pale cream to cream to brown
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Melting Point (clear melt)
138.0-145.0?C
Assay (HPLC)
≥98.5%
Identification (FTIR)
Conforms

Description

3-(2-Furyl)acrylic acid is a reactant used in the preparation of heteroaryl-substituted bis-trifluoromethyl carbinols and trifluoroacetophenone derivatives malonyl-CoA decarboxylase (MCD) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Furyl)acrylic acid is a reactant used in the preparation of heteroaryl-substituted bis-trifluoromethyl carbinols and trifluoroacetophenone derivatives malonyl-CoA decarboxylase (MCD) inhibitors.

Solubility
Soluble in dichloromethane and diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text