1,7-Dioxaspiro[5.5]undecane, 98%, Thermo Scientific Chemicals
1,7-Dioxaspiro[5.5]undecane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,7-Dioxaspiro[5.5]undecane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B21664.22
also known as B21664-22
Price (USD)/ Each
529.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
529.00
Add to cart
1,7-Dioxaspiro[5.5]undecane, 98%, Thermo Scientific Chemicals
Catalog numberB21664.22
Price (USD)/ Each
529.00
-
Add to cart
Chemical Identifiers
CAS180-84-7
IUPAC Name1,7-dioxaspiro[5.5]undecane
Molecular FormulaC9H16O2
InChI KeyGBBVHDGKDQAEOT-UHFFFAOYSA-N
SMILESC1CCC2(CCCCO2)OC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4605-1.4655 @ 20?C
FormLiquid
Suitable for use in organic farming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suitable for use in organic farming.

Solubility
Soluble in water 1.0g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Brett D Schwartz, et al. Spiroacetal biosynthesis: (+/-)-1,7-dioxaspiro[5.5]undecane in Bactrocera cacuminata and Bactrocera oleae (Olive Fruit Fly).Org. Lett.20057(6), 1173-1176.
  2. Kenji Mori and Hidenori Watanabe. Enhancement of optical purity of a product by removing unwanted diastereomers in the course of a chiral synthesis: new synthesis of the stereoisomers of 2,8-dimethyl-1,7-dioxaspiro[5.5]undecane, the pheromone of andrena wilkella.Tetrahedron.,198642(1), 295-304.