Thermo Scientific Chemicals

Calix[8]arene, 97%, Thermo Scientific Chemicals

Catalog number: B21666.03
1 g, Each
Thermo Scientific Chemicals

Calix[8]arene, 97%, Thermo Scientific Chemicals

Catalog number: B21666.03
1 g, Each
Quantity
Catalog number: B21666.03
also known as B21666-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
82452-93-5
IUPAC Name
nonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21(53),22,24,27(52),28,30,33(51),34,36,39(50),40,42,45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
Molecular Formula
C56H48O8
InChI Key
HDPRHRZFFPXZIL-UHFFFAOYSA-N
SMILES
OC1=C2CC3=CC=CC(CC4=CC=CC(CC5=CC=CC(CC6=C(O)C(CC7=C(O)C(CC8=C(O)C(CC9=C(O)C(CC1=CC=C2)=CC=C9)=CC=C8)=CC=C7)=CC=C6)=C5O)=C4O)=C3O
Appearance (Color)
White to cream
Form
Powder
Proton NMR
Conforms to structure
Purity (DSC)
>96.0 to <104.0%

Description

It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.

Solubility
Sparingly soluble in water (5.8E-6 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text