Thermo Scientific Chemicals

4-Bromo-2,6-dimethylphenol, 99%, Thermo Scientific Chemicals

Catalog number: B21669.22
100 g, Each
Thermo Scientific Chemicals

4-Bromo-2,6-dimethylphenol, 99%, Thermo Scientific Chemicals

Catalog number: B21669.22
100 g, Each
Quantity
Catalog number: B21669.22
also known as B21669-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2374-05-2
IUPAC Name
4-bromo-2,6-dimethylphenol
Molecular Formula
C8H9BrO
InChI Key
ZLVFYUORUHNMBO-UHFFFAOYSA-N
SMILES
CC1=CC(Br)=CC(C)=C1O
Appearance (Color)
Cream or red to brown
Form
Crystals or powder or crystalline powder or lumpy solid
Assay (GC)
≥96.0%
Melting Point
72-81°C

Description

It is used as a preservatives for coatings, slurries and to control microbial fouling in paper mills , oil field and leather process and water treatment process against microbial attack.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a preservatives for coatings, slurries and to control microbial fouling in paper mills , oil field and leather process and water treatment process against microbial attack.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text