Thermo Scientific Chemicals

15-Hydroxypentadecanoic acid, 99+%, Thermo Scientific Chemicals

Catalog number: B21670.06
5 g, Each
Thermo Scientific Chemicals

15-Hydroxypentadecanoic acid, 99+%, Thermo Scientific Chemicals

Catalog number: B21670.06
5 g, Each
Quantity
Catalog number: B21670.06
also known as B21670-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4617-33-8
IUPAC Name
15-hydroxypentadecanoate
Molecular Formula
C15H29O3
InChI Key
BZUNJUAMQZRJIP-UHFFFAOYSA-M
SMILES
OCCCCCCCCCCCCCCC([O-])=O
Appearance (Color)
White
Form
Powder or fused solid
Assay (Aqueous acid-base Titration)
≥99.0 to ≤101.0%
Assay (Silylated GC)
≥99.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text