Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-proline, 98+%, Thermo Scientific Chemicals

Catalog number: B21672.06
5 g, Each
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-proline, 98+%, Thermo Scientific Chemicals

Catalog number: B21672.06
5 g, Each
Quantity
Catalog number: B21672.06
also known as B21672-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1148-11-4
IUPAC Name
(2S)-1-[(benzyloxy)carbonyl]pyrrolidine-2-carboxylate
Molecular Formula
C13H14NO4
InChI Key
JXGVXCZADZNAMJ-NSHDSACASA-M
SMILES
[O-]C(=O)[C@@H]1CCCN1C(=O)OCC1=CC=CC=C1
Form
Powder or crystalline powder or crystals or granules
Optical Rotation
-58° to -62° (c=1 in acetic acid)
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

N-(Benzyloxycarbonyl)-L-proline is a potent inhibitor of prolidase; a specific peptidase that cleaves dipeptides with a C-terminal prolyl and hydroxylprolyl residue. It is also used in the synthesis of N-(L-Prolyl)-β-alanine which is a derivative of the naturally occurring beta amino acid β-Alanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Benzyloxycarbonyl)-L-proline is a potent inhibitor of prolidase; a specific peptidase that cleaves dipeptides with a C-terminal prolyl and hydroxylprolyl residue. It is also used in the synthesis of N-(L-Prolyl)-β-alanine which is a derivative of the naturally occurring beta amino acid β-Alanine.

Solubility
Solubility in methanol, almost transparency.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text