4?-Methylpropiophenone is a chemical reagent used in electrocarboxylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Methylpropiophenone is a chemical reagent used in electrocarboxylation reactions.
Solubility
Soluble in chloroform and hexane.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Shu-Feng Zhao; Mei-Xia Zhu; Kai Zhang; Huan Wang; Jia-Xing Lu. Alkaloid induced asymmetric electrocarboxylation of 4-methylpropiophenone. Tetrahedron Letters. 2011, 52 (21), 2702-2705.
- V. Karunakaran; V. Balachandran. Experimental and theoretical investigation of the molecular structure, conformational stability, hyperpolarizability, electrostatic potential, thermodynamic properties and NMR spectra of pharmaceutical important molecule: 4'-Methylpropiophenone. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy, 2014, 128(2), 1-14.