It is used in the synthesis of silyl and stannyl pyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis of silyl and stannyl pyrimidines.
Solubility
Soluble in chloroform, ethyl acetate and methanol.
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- Cyril Párkányi, et al. Synthesis of silyl and stannyl pyrimidines.J. Organomet. Chem.,1988342(1), 1-7.
- P. J. Bray, et al. Pure Quadrupole Resonance Determinations of Molecular Charge Distributions. I. Substituted Quinones, Quinolines, Pyridines, Pyrimidines, and Triazines.J. Chem. Phys.,19582899.