Thermo Scientific Chemicals

Octafluoronaphthalene, 96%, Thermo Scientific Chemicals

Catalog number: B21699.14
25 g, Each
Thermo Scientific Chemicals

Octafluoronaphthalene, 96%, Thermo Scientific Chemicals

Catalog number: B21699.14
25 g, Each
Quantity
Catalog number: B21699.14
also known as B21699-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
313-72-4
IUPAC Name
octafluoronaphthalene
Molecular Formula
C10F8
InChI Key
JDCMOHAFGDQQJX-UHFFFAOYSA-N
SMILES
FC1=C(F)C2=C(F)C(F)=C(F)C(F)=C2C(F)=C1F
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or lumps
Identification (FTIR)
Conforms
Melting Point (clear melt)
82.0-91.0?C
Assay (GC)
≥95.0%

Description

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water. Solubility in toluene, almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text