Thermo Scientific Chemicals

2-Bromo-4-methylphenol, 97%, Thermo Scientific Chemicals

Catalog number: B21701.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-4-methylphenol, 97%, Thermo Scientific Chemicals

Catalog number: B21701.06
5 g, Each
Quantity
Catalog number: B21701.06
also known as B21701-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6627-55-0
IUPAC Name
2-bromo-4-methylphenol
Molecular Formula
C7H7BrO
InChI Key
MTIDYGLTAOZOGU-UHFFFAOYSA-N
SMILES
CC1=CC=C(O)C(Br)=C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.5770-1.5820 @ 20?C

Description

It is applied as a compound responsible for iodine off-flavor in wines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a compound responsible for iodine off-flavor in wines.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text