Thermo Scientific Chemicals

3-Hydroxybenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: B21702.14
25 g, Each
Thermo Scientific Chemicals

3-Hydroxybenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: B21702.14
25 g, Each
Quantity
Catalog number: B21702.14
also known as B21702-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
873-62-1
IUPAC Name
3-hydroxybenzonitrile
Molecular Formula
C7H5NO
InChI Key
SGHBRHKBCLLVCI-UHFFFAOYSA-N
SMILES
OC1=CC=CC(=C1)C#N
Appearance (Color)
Pale brown
Form
Powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
76.0-83.0?C

Description

3-Hydroxybenzonitrile, is an intermediate that can be used for the synthesis of new Serotonin 5-HT1A receptor agonists having antinociceptive activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzonitrile, is an intermediate that can be used for the synthesis of new Serotonin 5-HT1A receptor agonists having antinociceptive activity.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text