Thermo Scientific Chemicals

2-Propionylthiazole, 99%, Thermo Scientific Chemicals

Catalog number: B21711.09
10 g, Each
Thermo Scientific Chemicals

2-Propionylthiazole, 99%, Thermo Scientific Chemicals

Catalog number: B21711.09
10 g, Each
Quantity
Catalog number: B21711.09
also known as B21711-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
43039-98-1
IUPAC Name
1-(1,3-thiazol-2-yl)propan-1-one
Molecular Formula
C6H7NOS
InChI Key
TYRAENAWSLPSLW-UHFFFAOYSA-N
SMILES
CCC(=O)C1=NC=CS1
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5355-1.5385 @ 20?C
Appearance (Color)
Clear colorless

Description

Used as flavoring agent and fragrance chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as flavoring agent and fragrance chemical compounds.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text