4-Methyl-5-vinylthiazole, 98+%, Thermo Scientific Chemicals
4-Methyl-5-vinylthiazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methyl-5-vinylthiazole, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21719.06
also known as B21719-06
Price (USD)/ Each
37.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
37.10
Add to cart
4-Methyl-5-vinylthiazole, 98+%, Thermo Scientific Chemicals
Catalog numberB21719.06
Price (USD)/ Each
37.10
-
Add to cart
Chemical Identifiers
CAS1759-28-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
FormLiquid
Refractive Index1.5655-1.5695 @ 20?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only